Filter discs, with hole, VitraPOR®
Supplier: ROBU GLASFILTER
17020 17021 17022 17023 17024 17029 17003 17009 17004 17000 17001 17002
ROBU17020EA 69 GBP
ROBU17020 ROBU17021 ROBU17022 ROBU17023 ROBU17024 ROBU17029 ROBU17003 ROBU17009 ROBU17004 ROBU17000 ROBU17001 ROBU17002
Filter discs, with hole, VitraPOR®
Filters Filter Discs
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
  • For gas distribution tubes
Made of borosilicate glass. Glass filters discs are divided into different porosity classes.
Order Now
SPECIFICATIONS
Environmentally Preferable