Utility clamps, three pronged
Supplier: R L ENTERPRISES
1029
RLAA1029EA 134 GBP
RLAA1029
Utility clamps, three pronged
Clamps General Purpose Clamps
This 3-pronged clamp made of diecast aluminium alloy features plastic coated jaws.
  • With screwed shaft and nickel-plated bosshead
  • Plastic coated jaws are chemical resistant
  • Can be screwed up and down shaft 35 mm and locked in any position with a wing nut
The bosshead accepts rods up to 13 mm, which are secured with a stainless steel thumb screw.