dNTP mix, 2 mM
Supplier: Thermo Fisher Scientific
R0242
R0242EA 130 GBP
R0242
dNTP mix, 2 mM
Nucleic Acid Reagents Nucleotides
dNTP mix contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP, each at a final concentration of 2 mM.
  • Greater than 99% purity confirmed by HPLC
  • Free of human and E. coli DNA
  • Highly stable
Suitable for used in all molecular biology applications, including PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, RDA, MDA, DNA labelling, and DNA sequencing.