Dissolution vessels, QLA
Supplier: QUALITY LAB ACCESSORIES
GLA02K-01 GLA02K-DK GLA02K-EW GLA500-01 GLA500-DK GLA500-HR GLA900-01 GLA900-AD GLA900-AH GLA900-AV GLA900-CA GLA900-DK GLA900-EW GLA900-HR GLA900-PT GLA900-ST GLA900-VK GLA9CR-AD GLA9CR-DK GLA9PK-01 PLA900-01 PLA900-AD PLA900-AH PLA900-AV PLA900-DK PLA900-EW PLA900-HR GLA9AM-VK
QLAAGLA02K-01EA 506 GBP
QLAAGLA02K-01 QLAAGLA02K-DK QLAAGLA02K-EW QLAAGLA500-01 QLAAGLA500-DK QLAAGLA500-HR QLAAGLA900-01 QLAAGLA900-AD QLAAGLA900-AH QLAAGLA900-AV QLAAGLA900-CA QLAAGLA900-DK QLAAGLA900-EW QLAAGLA900-HR QLAAGLA900-PT QLAAGLA900-ST QLAAGLA900-VK QLAAGLA9CR-AD QLAAGLA9CR-DK QLAAGLA9PK-01 QLAAPLA900-01 QLAAPLA900-AD QLAAPLA900-AH QLAAPLA900-AV QLAAPLA900-DK QLAAPLA900-EW QLAAPLA900-HR QLAAGLA9AM-VK
Dissolution vessels, QLA
Dissolution Apparatus
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
  • Made of high quality borosilicate glass or plastic
  • Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Order Now
SPECIFICATIONS
Environmentally Preferable