Dissolution vessel covers
Supplier: QUALITY LAB ACCESSORIES
COVERD-00 COVERD-01 COVERD-02 COVERD-HG COVERH-00 COVERH-01 COVERH-02 COVERH-03 COVERH-04 COVERH-HG COVERH-LL COVERV-01 COVERV-02 COVERV-03 COVERV-04 COVERV-6K COVERV-HG COVERV-LL COVPLG-7D COVPLG-DK
QLAACOVERD-00EA 213 GBP
QLAACOVERD-00 QLAACOVERD-01 QLAACOVERD-02 QLAACOVERD-HG QLAACOVERH-00 QLAACOVERH-01 QLAACOVERH-02 QLAACOVERH-03 QLAACOVERH-04 QLAACOVERH-HG QLAACOVERH-LL QLAACOVERV-01 QLAACOVERV-02 QLAACOVERV-03 QLAACOVERV-04 QLAACOVERV-6K QLAACOVERV-HG QLAACOVERV-LL QLAACOVPLG-7D QLAACOVPLG-DK
Dissolution vessel covers
Lids Reaction Vessel Lids
Standard and low loss vessel covers for most dissolution baths.
  • Slots and holes for temperature verification, sample introduction and sample collection should be closed when not in use
  • Condensation is reduced by using vessel covers with conical bottoms that allow condensation to return back to vessel
The loss of media due to evaporation can be considerable. Vesssel covers reduce these issues and provide openings for cannula, cannula stoppers, temperature verification, and sample introduction. There are many varieties of covers to choose from,More Product Information