Face Shield Plus
Supplier: Portwest
PW96CLR
PRTWPW96CLREA 26.7 GBP
PRTWPW96CLR
Face Shield Plus
Face Shields
This face shield protects against molten metal splashes and hot solid projections.
  • Browguard including clear polycarbonate visor with aluminum edge
  • Protects from impacts, metal splashes, and hot solid projections
  • Suitable for a wide range of applications
  • Wheel ratchet size adjustment for easy fitting
  • Simple and secure visor replacement
  • Retail bag aids presentation for retail sales
Superior front and side face protection. Ratchet adjustment for comfort fit. Replacement visor available (PW99).
Certifications: ANSI/ISEA Z87 +.