Browguard with mesh visor, PW93
Supplier: Portwest
PW93BKR
PRTWPW93BKREA 20.4 GBP
PRTWPW93BKR
Browguard with mesh visor, PW93
Face Shields
Mesh visor with PP browguard, suitable for protection against large highspeed particles. The ratchet adjustment system is designed for easy use.
  • Wheel ratchet size adjustment for easy fitting
  • Soft foam sweat band included
  • Can be adapted to be used over prescription glasses
  • Individually packed for vending machines
Materials: galvanised carbon steel, polypropylene.
Certifications: EN 1731 (F); EN 166 (B); CE