Cleanroom face masks, Auckland, P410
Supplier: Portwest
P410BLUL P410BLUM P410BLUS
PRTWP410BLULEA 15.3 GBP
PRTWP410BLUL PRTWP410BLUM PRTWP410BLUS
Cleanroom face masks, Auckland, P410
Masks Cleanroom Masks
Economical half mask with low profile design providing a large field of vision.
  • The soft adjustable strap ensures perfect fit to wide face contours
  • Equipped with a dual (screw in) filter for enhanced breathing comfort
  • Sweat port on the face piece for hot and humid environments
Manufactured from a durable yet soft non allergic TPE material which provides an excellent fit.
Certifications: EN 140
Delivery information: Filters sold separately.