Palladium standard solution, for ICP, in 5% HNO₃, VHG
Supplier: LGC Standards PROMOCHEM
VHG-PPDN-100 VHG-PPDN-50 VHG-PPDN-500 VHG-TPDN-100 VHG-TPDN-500
PROOVHG-PPDN-100EA 94.6 GBP
PROOVHG-PPDN-100 PROOVHG-PPDN-50 PROOVHG-PPDN-500 PROOVHG-TPDN-100 PROOVHG-TPDN-500
Palladium standard solution, for ICP, in 5% HNO₃, VHG
Standards Cationic Standards Palladium Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.
Order Now
SPECIFICATIONS
Environmentally Preferable