Dieldrin

Supplier: LGC Standards PROMOCHEM
Danger

NMIAP1747
PROONMIAP1747EA 327 GBP
PROONMIAP1747
Dieldrin
Dieldrin
Formula: C₁₂H₈Cl₆O
MW: 380.91 g/mol
Boiling Pt: 385
Melting Pt: 174…175 °C
Density: 1.75 g/cm³ (20 °C)
CAS Number: 60-57-1
EINECS: 200-484-5
UN: 2761
ADR: 6.1 II
Merck Index: 12,03152

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR