Industrial sludge - leachable and total metals, NIST-2782
Supplier: LGC Standards PROMOCHEM
NIST-2782
PROONIST-2782EA 1660 GBP
PROONIST-2782
Industrial sludge - leachable and total metals, NIST-2782
Standards Environmental Analysis Standards
This standard reference material is intended primarily for use in the evaluation of methods used for the analysis of sludges and other materials of a similar matrix.
A unit consists of 70 g of dried, pulverised and radiation sterilised industrial sludge.
Certifications: Traceable to NIST and SRM numbers.