Barbituric acid

Supplier: LGC Standards PROMOCHEM

Synonyms: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine

MM0265.03
PROOMM0265.03EA 610 GBP
PROOMM0265.03
Barbituric acid
Barbituric acid
Formula: C₄H₄N₂O₃
MW: 128.09 g/mol
Boiling Pt: 441
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00006666
CAS Number: 67-52-7
EINECS: 200-658-0
Merck Index: 13,00966

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR