Cyclopentane

Supplier: LGC Standards PROMOCHEM
Danger

Synonyms: Pentamethylene

CHE142
PROOCHE142EA 268 GBP
PROOCHE142
Cyclopentane
Cyclopentane
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 49
Melting Pt: –93.3 °C
Density: 0.746 g/cm³ (20 °C)
Flash Pt: –41 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
EINECS: 206-016-6
UN: 1146
ADR: 3 II
Merck Index: 13,02769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dd230057521cc77c90840c7adfece9c74c0a3b1&vid=352021851b7701a448c5b11d43e94d925d03f802&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR