3-Methylpentane

Supplier: LGC Standards PROMOCHEM
Danger

CHE120
PROOCHE120EA 268 GBP
PROOCHE120
3-Methylpentane
3-Methylpentane
Formula: C₆H₁₄
MW: 86.18 g/mol
Boiling Pt: 64
Melting Pt: –118 °C
Density: 0.664 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009342
CAS Number: 96-14-0
EINECS: 202-481-4
UN: 1208
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR