Progesterone

Supplier: LGC Standards PROMOCHEM

BP449
PROOBP449EA 390 GBP
PROOBP449
Progesterone
Progesterone
Formula: C₂₁H₃₀O₂
MDL Number: MFCD00003658
CAS Number: 57-83-0
EINECS: 200-350-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR