Oversized Rayon OB/GYN Swabs
Supplier: PURITAN MEDICAL
25-8081PR
PMPC25-8081PREA 546 GBP
PMPC25-8081PR
Oversized Rayon OB/GYN Swabs
Swabs Swabs for Sampling
Extra-absorbent oversized rayon tipped applicator with plastic handle.
  • Extra-absorbent large oversized tip
  • Long PS handles
  • Medical grade quality, sterile
Ideal for gathering specimens as well as OB/GYN preparation and examinations.
Packaging: 10 boxes of 50 wrapped packages.