Desalting columns, Zeba™, Pierce™
Supplier: Thermo Fisher Scientific
89877 89882 89890 89894 89891 89893 89889 89892 89878 89883 87773
PIER89877EA 207 GBP
PIER89877 PIER89882 PIER89890 PIER89894 PIER89891 PIER89893 PIER89889 PIER89892 PIER89878 PIER89883 PIER87773
Desalting columns, Zeba™, Pierce™
Chromatography Columns
These spin desalting columns are made of low protein binding PP and are compatible with a wide range of standard laboratory instruments and consumables. They provide >95% retention of salts and other small molecules.
  • Higher protein recovery
  • Minimal sample dilution
  • No screening fractions for protein or waiting for protein to emerge by gravity-flow
  • Easy-to-use with no cumbersome column preparation or equilibration
The Zeba 7K MWCO is recommended for processing proteins >7 kDa, and the Zeba 40K MWCO for processing proteins >40 kDa.