Spin filter plates, detergent removal, 96-well, Pierce™
Supplier: Thermo Fisher Scientific
88304
PIER88304EA 809 GBP
PIER88304
Spin filter plates, detergent removal, 96-well, Pierce™
Microplates Filter Plates
Pierce™ Detergent removal filter spin plates enable multiple-sample processing using a high-performance resin that offers effective detergent removal and excellent protein/peptide recovery. Pierce™ Detergent removal resin efficiently binds to and removes high concentrations of detergents from 10 µl to 1 ml samples with minimal sample loss.
  • Fast and efficient protocol
  • Removes greater than 95% of detergents with high protein recovery
  • Helps improves mass spectrometry peptide coverage
  • Best for processing samples with proteins or peptides at greater than 100 µg/
More Product Information