Electrophoresis running buffers, Pierce™
Supplier: Thermo Fisher Scientific
28362
PIER28362EA 94.6 GBP
PIER28362
Electrophoresis running buffers, Pierce™
Buffers Electrophoresis Buffers Running Buffers
These are space-saving stock solution that is ideal for quickly preparing running buffer used for gel electrophoresis.
  • Eliminates the possibility of powder remaining in a packet
  • Eliminates time spent waiting for powder to dissolve
  • Storage as concentrated stock minimises bench space needed for solutions
Tris-HEPES-SDS buffer - when diluted 20-fold in water, the solution yields 0,1 M Tris, 0,1 M HEPES, 3 mM SDS, pH 8
Tris-glycine-SDS buffer - when diluted 10-fold in water, the solution yields 0,025 M Tris, 0,192 M glycine, 0,1% SDS, pH 8,5