Pegylation reagents, Methyl-PEG-NHS-Esters, Pierce™
Supplier: Thermo Fisher Scientific
22341 22509 22685 22342 22424 22686 22687
PIER22341EA 297 GBP
PIER22341 PIER22509 PIER22685 PIER22342 PIER22424 PIER22686 PIER22687
Pegylation reagents, Methyl-PEG-NHS-Esters, Pierce™
Protein Purification Protein Purification Reagents
Pierce™ MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) and TMS(PEG)12 is a three-branched, polyethylene glycol compound (3× 12 PEG units). Both are activated as NHS esters for covalent pegylation of primary amines on proteins (eg. lysines) or assay surfaces.
  • Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimised for a specific application
  • PEG spacer provides advantages that include increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • NHS-activated for efficient PEGylation of primary amines at pH 7 to 9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
  • Easy to follow instructions increase the likelihood of a successful outcome
MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript 'n' denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH₂), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.
TMS(PEG)12 is the abbreviation for a branched trimethyl (TM) and succinimide ester (S) derivative of polyethylene glycol (PEG) for efficient and specific modification of primary amines. Each methyl-terminated PEG (mPEG) branch contains 12 ethylene glycol units. The three branches are attached to a 4-unit PEG stem that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at the distal end. The NHS ester is spontaneously reactive with primary amines (—NH₂), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.