DL-α-Tocopherol acetate, Supelco®

Supplier: Merck

Synonyms: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate

PHR1030-500MG
PHR1030-500MGEA 85 GBP
PHR1030-500MG
DL-α-Tocopherol acetate, Supelco®
DL-α-Tocopherol acetate
Formula: C₃₁H₅₂O₃
MW: 472.75 g/mol
MDL Number: MFCD00072042
CAS Number: 7695-91-2
EINECS: 231-710-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2871ffba58af9ce8f5346e92939b2433373c6013&vid=2871ffba58af9ce8f5346e92939b2433373c6013&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR