LC-MS nitrogen generators
Supplier: PerkinElmer
N9306211 N9306214
PERCN9306211EA 43660 GBP
PERCN9306211 PERCN9306214
LC-MS nitrogen generators
Gas Generators Nitrogen Generators
Nitrogen generators are designed to supply single or multiple LC-MS instruments with dry nitrogen at purities of 98 – 99,5%. The generator can also be used for solvent evaporation as well as supplying dry nitrogen to analytical instruments.
Order Now
SPECIFICATIONS
Environmentally Preferable