Column cutter
Supplier: PerkinElmer
N9301376
PERCN9301376EA 152 GBP
PERCN9301376
Column cutter
Cutters
Ceramic wafer scribe is inexpensive and ideal for cutting polyimide fused silica capillary columns and guard columns.
  • Easy to hold
  • All four sides can be used as a cutting tool