Tin capsule kit, large
Supplier: PerkinElmer
N2411362
PERCN2411362EA 267 GBP
PERCN2411362
Tin capsule kit, large
Sample Containers General Purpose Containers
Large tin capsule used for the analysis of nonvolatile solid samples. It is mostly used in the combustion of the sample.
Tin capsules are routinely used to analyse polymers, soils, coal, dry dairy products, plant leaves, paper, textiles, calibration standards, and many organic and pharmaceutical products.