Phenethylbiguanide hydrochloride, Supelco®

Supplier: Merck

Synonyms: Phenformin hydrochloride, 1-Carbamimidamido-N-(2-phenylethyl)methanimidamide Hydrochloride

P7045-10G P7045-1G
P7045-10GEA 129 GBP
P7045-10G P7045-1G
Phenethylbiguanide hydrochloride, Supelco®
Phenethylbiguanide hydrochloride
Formula: C₁₀H₁₆ClN₅
MW: 241.7205 g/mol
MDL Number: MFCD00035039
CAS Number: 834-28-6
EINECS: 212-637-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24e809c84160e2b1cd4156bbf436e2150c32b242&vid=62cc3d474d808754c6df98d06ea9aacf26b260d2&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3013150b096cfa79efdba884237debbec668b887&vid=c4d8fdca7c0a1656c680da0da3342bebaba5b751&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR