L(+)-Glutamine, powder, non_sterile

Supplier: BIOWEST

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

P1012-500GR P1012-1KG
P1012-500GREA 0 GBP
P1012-500GR P1012-1KG
L(+)-Glutamine, powder, non_sterile
L(+)-Glutamine
Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Boiling Pt: 355
Melting Pt: 185…186 °C
Density: 1.5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR