Transduction reagent, ViroMag™
Supplier: OZ BIOSCIENCES
VM40100 KC30500 KC30600 VM40200 VM41000
OZBIVM40100EA 99.1 GBP
OZBIVM40100 OZBIKC30500 OZBIKC30600 OZBIVM40200 OZBIVM41000
Transduction reagent, ViroMag™
Transfection Reagents
ViroMag™ transduction reagent is a magnetic nanoparticles formulation dedicated to increase virus infection and transduction capacities. Based on Magnetofection™ technology, this reagent allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus transduction with extremely low vector doses. Due to its specific properties, ViroMag™ is ideal to infect non permissive cells.
  • Increases transduction efficiency
  • Accelerates the transduction process
  • Efficient for hard-to-infect and non permissive cells
  • Concentrates the viral dose onto the cells
  • Suitable for all type of virus
  • Synchronise cells adsorption/infection without modifications of the virus
This reagent demonstrates an exceptionally high efficiency to promote, control and assist viral transductions so that no molecular biology processes or biochemical modifications are required.
Store at +4 °C.
Ordering information: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Delivery information: Shipped at room temperature.
Order Now
SPECIFICATIONS
Cell specific Mammalian cells
Environmentally Preferable