Transfection control plasmids, pVectOZ
Supplier: OZ BIOSCIENCES
PL00020 PL00120 PL00010 PL00110 PL00030 PL00130 PL00040 PL00140 PL00050 PL00150
OZBIPL00020EA 335 GBP
OZBIPL00020 OZBIPL00120 OZBIPL00010 OZBIPL00110 OZBIPL00030 OZBIPL00130 OZBIPL00040 OZBIPL00140 OZBIPL00050 OZBIPL00150
Transfection control plasmids, pVectOZ
Vectors, Plasmids and Libraries
pVectOZ transfection plasmids are engineered in an optimised backbone and are ideal for all transfections.
  • Highest levels of transgene expression in mammalian cells and tissues
  • Suitable for all transfection applications (in vivo and in vitro)
  • High copy number production in Escherichia coli
  • LPS-endotoxin free, supercoiled and highly purified: transfection grade approved
  • Recommended for positive control and optimisation of all transfection experiments
Comprising five expression vectors encoding for the most popular reporter genes (CAT, GFP, LacZ, Luciferase, SEAP).
Delivery information: Shipped at room temperature
Packaging: Store at –20 °C.