siRNA transfection reagent, Lullaby™
Supplier: OZ BIOSCIENCES
LL70500 LL71000 LL73000
OZBILL70500EA 201 GBP
OZBILL70500 OZBILL71000 OZBILL73000
siRNA transfection reagent, Lullaby™
Transfection Reagents
Lullaby™ is an ideal siRNA transfection reagent for gene silencing. Based on the TEE-technology, it has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and very low toxicity. RNA interference is a powerful technique to shut down genes expression in cells and organisms. This silencing effect constitutes a very helpful tool to study gene function and a promising approach for new therapeutic treatments.
  • Effective with very low doses of siRNA
  • Minimised off-target effects
  • Suitable for siRNA, shRNA, dsRNA and more
  • Applicable to a broad range of cells
  • Serum compatible and non-toxic
  • Straightforward protocol
Lullaby™ transfection reagent introduces siRNA duplexes in a variety of cells with a very high efficiency leading to exceptional knockdown effects with low doses of siRNA.
Store at +4 °C.
Delivery information: Shipped at room temperature.
Order Now
SPECIFICATIONS
Environmentally Preferable
dsRNA Yes
High throughput screening Yes
shRNA Yes
siRNA Yes