DNA transfection reagent, HYPE-CHO™
Supplier: OZ BIOSCIENCES
HYC01500 HYC03000 HYC15000 HYC30000
OZBIHYC01500EA 212 GBP
OZBIHYC01500 OZBIHYC03000 OZBIHYC15000 OZBIHYC30000
DNA transfection reagent, HYPE-CHO™
Transfection Reagents
HYPE-CHO™ transfection kit has been designed for large scale up transient transfection and high protein expression, such as antibodies. The system is optimised for maximum efficiency in all CHO-S cells.
  • Reach optimal protein and antibody production yield
  • Suitable for all CHO cells growing in suspension
  • Compatible for any synthetic or regular media used for protein production
  • Ideal for bioreactor, spinner or flasks
  • Animal origin free
  • Suitable for bioproduction and biopharmaceutical manufacturing
HYPE-CHO™ has been used and validated with cells from different origins (CHO, CHO-S, rCHO or any CHO-related cells) cultured in suspension (flask, spinner and bioreactor) to produce proteins.
Delivery information: Shipped at room temperature.
Caution: Store Hype-CHO reagent and BCHO at +4 °C.