DNA transfection reagent, HYPE-293™
Supplier: OZ BIOSCIENCES
HY29315 HY29330 HY293150 HY293300
OZBIHY29315EA 212 GBP
OZBIHY29315 OZBIHY29330 OZBIHY293150 OZBIHY293300
DNA transfection reagent, HYPE-293™
Transfection Reagents
HYPE-293™ transfection kits are dedicated to achieving high yield protein expression in HEK-293, 293-S, 293-F, or any 293-related cells grown in suspension.
  • For optimal protein and antibody production yield
  • Suitable for all HEK293 cells grown in suspension
  • Compatible with any synthetic or regular media used for protein production
  • Ideal for bioreactor, spinner, or flasks
  • Animal-free origin
Scale up to larger volumes for production of milligrams of protein per litre of cell culture is straightforward, with simple and cost efficient handling steps.
Store HYPE-293 reagent and B293 reagent at –20 °C.
Delivery information: Shipped at room temperature.