Accessories for Anaerobic Systems, Oxoid
Supplier: THERMO FISHER DIAGNOSTICS
AG0027A AG0029A AG0030A AN0010C AN0025A AN0035A BR0055B HP0016A HP0017A HP0018A HP0020A HP0026A HP0053A AG0020C CD0025A AN0005C AN0020C AG0031A AG0060C CD0020C
OXOIAG0027AEA 0 GBP
OXOIAG0027A OXOIAG0029A OXOIAG0030A OXOIAN0010C OXOIAN0025A OXOIAN0035A OXOIBR0055B OXOIHP0016A OXOIHP0017A OXOIHP0018A OXOIHP0020A OXOIHP0026A OXOIHP0053A OXOIAG0020C OXOICD0025A OXOIAN0005C OXOIAN0020C OXOIAG0031A OXOIAG0060C OXOICD0020C
Accessories for Anaerobic Systems, Oxoid
Anaerobic Systems
Where an AnaeroGen sachet is placed in a sealed jar, the atmospheric oxygen in the jar is rapidly absorbed with the simultaneous generation of carbon dioxide. This novel method differs from others commonly used in that the reaction proceeds with no evolution of hydrogen, and therefore, does not require a catalyst. Furthermore, water is not required to activate the reaction. When used as directed, the AnaeroGen sachet will reduce the oxygen level in the jar to below 1% within 30 minutes. The resulting carbon dioxide level will be between 9% and 13%.
CampyGen Compact for 1 or 2 Petri dishes, isMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable