Omnifit® EZ SolventPlus™ Glass Chromatography Columns for FPLC and LPLC
Supplier: OMNIFIT
006EZS-06-05AF 006EZS-06-25AF 006EZS-06-05AA 006EZS-10-10AA 006EZS-10-15AF 006EZS-10-15FF 006EZS-35-15AF 006EZS-10-10AF 006EZS-15-15AF 006EZS-25-25AF 006EZS-25-50AA 006EZS-25-50AF 006EZS-25-50FF 006EZS-10-33AA 006EZS-15-33AA 006EZS-25-33AA 006EZS-50-25AF 006EZS-50-25AA
OMNI006EZS-06-05AFEA 1290 GBP
OMNI006EZS-06-05AF OMNI006EZS-06-25AF OMNI006EZS-06-05AA OMNI006EZS-10-10AA OMNI006EZS-10-15AF OMNI006EZS-10-15FF OMNI006EZS-35-15AF OMNI006EZS-10-10AF OMNI006EZS-15-15AF OMNI006EZS-25-25AF OMNI006EZS-25-50AA OMNI006EZS-25-50AF OMNI006EZS-25-50FF OMNI006EZS-10-33AA OMNI006EZS-15-33AA OMNI006EZS-25-33AA OMNI006EZS-50-25AF OMNI006EZS-50-25AA
Omnifit® EZ SolventPlus™ Glass Chromatography Columns for FPLC and LPLC
Chromatography Columns
EZ SolventPlus™ columns are designed for maximum chemical compatibility. Borosilicate glass and PTFE construction, with Chemraz® o-rings and PTFE frits, only the most inert materials are present in the fluid path. ETFE end caps and fitting nuts ensure a chemically inert connection into and out of the system.
  • Available in six diameters and a variety of lengths
  • Operating temperature: 4 to 40 °C
  • Glass column is made up of borosilicate glass
Designed for techniques using organic solvents such as reversed phase chromatography, but equally compatible withMore Product Information