Viral DNA/RNA Xpress kit, CE IVD, Mag-Bind®
Supplier: OMEGA BIO-TEK
M62192304CEIVD
OMEGM62192304CEIVDEA 4550 GBP
OMEGM62192304CEIVD
Viral DNA/RNA Xpress kit, CE IVD, Mag-Bind®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Mag-Bind® Viral DNA/RNA Xpress Kit CE IVD follows a magnetic bead-based approach for the rapid and reliable isolation of viral DNA and RNA from nasopharyngeal (NP) swab specimens that are dry or in viral transport media (VTM), from saliva and other sample sources.
Mag-Bind® Viral DNA/RNA Xpress Kit CE IVD follows a magnetic bead-based approach for the rapid and reliable isolation of viral DNA and RNA from nasopharyngeal (NP) swab specimens that are dry or in viral transport media (VTM), from saliva and other sample sources. The extraction methodology is easily adaptable to various automated systems and can also be scaled up or down depending on the amount of starting sample amount used. The kit utilizes the proven Mag-Bind® technology that enables the purification of high-quality nucleic acids that are free of proteins, nucleases, and other impurities. The purified nucleic acids are ready for direct use in downstream applications such as qPCR, RT-qPCR, and more.
Certifications: CE-IVD IVDR