Anti-MAS1 Rabbit Polyclonal Antibody (DyLight® 405)

Supplier: Novus Biologicals
NLS1531V
NOVUNLS1531VEA 667 GBP
NOVUNLS1531V
Anti-MAS1 Rabbit Polyclonal Antibody (DyLight® 405)
Antibodies
Type: Primary
Antigen: MAS1 (MAS1 oncogene)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol MAS1
Environmentally Preferable
Antigen name MAS1 proto-oncogene, G protein-coupled receptor
Antigen synonyms MGRA|MAS
Antibody type Primary
Clonality Polyclonal
Conjugation DyLight® 405
Reactivity Human,Mouse
Host Rabbit
Gene ID 4142
Isotype
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR