Anti-HSPA13 Rabbit Polyclonal Antibody (DyLight® 650)

Supplier: Novus Biologicals
NBP1-77455C
NOVUNBP1-77455CEA 650 GBP
NOVUNBP1-77455C
Anti-HSPA13 Rabbit Polyclonal Antibody (DyLight® 650)
Antibodies
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR