Chromata ChIP Histone H3 [Monomethyl Lys9] Kit

Supplier: Novus Biologicals

NBP1-71713
NOVUNBP1-71713EA 803 GBP
NOVUNBP1-71713
Chromata ChIP Histone H3 [Monomethyl Lys9] Kit
Assays Immunological Assays

Chromata ChIP Histone H3 [Monomethyl Lys9] Kit


Tested Applications: Chromatin Immunoprecipitation.

Order Now

SPECIFICATIONS

Antigen symbol Chromata ChIP Histone H3
Environmentally Preferable
ChIP Yes
Product type KIT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR