Anti-ATF3 Rabbit Polyclonal Antibody (DyLight® 405)

Supplier: Novus Biologicals
NBP1-02935V
NOVUNBP1-02935VEA 703 GBP
NOVUNBP1-02935V
Anti-ATF3 Rabbit Polyclonal Antibody (DyLight® 405)
Antibodies
Type: Primary
Antigen: ATF3 (activating transcription factor 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol ATF3
Antigen name Activating transcription factor 3
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation DyLight® 405
Reactivity
Host Rabbit
Gene ID 467
Isotype
Western blot Yes
ELISA Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR