Modular deck system, PIG®
Supplier: NEW PIG
PAK524 PAK529 PAK531 PAK538 PAK726-BK
NEWPPAK524EA 249 GBP
NEWPPAK524 NEWPPAK529 NEWPPAK531 NEWPPAK538 NEWPPAK726-BK
Modular deck system, PIG®
Spill Control Materials
Modular deck system ideal to create a customised drum storage area that shares sump capacity. With flow-through sump design liquid passes freely from one sump to another.
  • Linear LDPE sump with injection molded PE grate
  • Low-profile design is easy to load or unload
  • Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe
  • LDPE construction resists UV rays, rust, corrosion and most chemicals
  • Textured grating adds traction and lifts out for easy sump access
  • Translucent white sidewalls allow easy visual inspection