Waste Container
Supplier: NEW PIG
NPK201-999
NEWPNPK201-999EA 78.9 GBP
NEWPNPK201-999
Waste Container
Bins Waste Bins
This 2-wheeled bin is ideal for many different types of waste collection like paper and cardboard, glass, packaging, organic materials, as well as being a robust container for spill control absorbents and accessories.
  • High-density polyethylene stabilised resistant to atmospheric conditions and combined effects of UV radiation and water for a long usable life
  • Great stability against fluctuations in temperature and total corrosion resistance
  • Manufactured using environmentally-friendly recyclable materials with no heavy metals in the pigments used
  • DIN
More Product Information