Silicone Hose
Supplier: NEUBERT VOLUME GLASSWAERE
6200-070-020 6200-090-020 6200-090-015 6201-060-030
NEUB6200-070-020EA 14.8 GBP
NEUB6200-070-020 NEUB6200-090-020 NEUB6200-090-015 NEUB6201-060-030
Silicone Hose
Tubing Transfer Tubing
Hose made of quality silicone rubber (VMQ).
  • Hardness 55 Shore A
  • Food safe
  • Peristaltic pumps suitable
Temperature resistant from –60 to +200 °C, short-term up to 260 °C.