Solid Glass Stopper
Supplier: NEUBERT VOLUME GLASSWAERE
1641-91-3435
NEUB1641-91-3435EA 90.3 GBP
NEUB1641-91-3435
Solid Glass Stopper
Stoppers Glass Stoppers
Six sided, solid glass stopper.
  • Made from Borosilicate glass 3.3
  • Six point handle