Anti-ANK3 Mouse Monoclonal Antibody

Supplier: Merck Millipore (Oncogene)
NB20-100
NB20-100EA 746 GBP
NB20-100
Anti-ANK3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ANK3 (ankyrin 3, node of Ranvier (ankyrin G))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR