Naphthol AS-GR phosphate disodium salt

Supplier: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

N3625-1G
N3625-1GEA 949 GBP
N3625-1G
Naphthol AS-GR phosphate disodium salt
Naphthol AS-GR phosphate disodium salt

Produces highly insoluble dark green stain in histochemical studies.

Formula: C₂₂H₁₆NO₅PNa₂
MW: 451.32 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00069449
CAS Number: 100929-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72cea00ddaca67c608823a5959df5aeb7163ffbb&vid=a325e7886f00cf927e143e1452d753ecd04ea0ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear to slightly hazy dark yellow solution at 50 mg/ml of water
Solubility Conforms to structure
IR Spectrum < or = 23% (7 Mol/Mol)
Water by Karl Fischer 6.6 to 7.1% Phosphorus
ICP-Atomic Emission Jun-03
Purity by thin layer chromatography * Anhydrous Basis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR