Hermetic Box, PP
Supplier: MULTIROIR
1368
MULC1368EA 194 GBP
MULC1368
Hermetic Box, PP
Boxes Storage Boxes
Certified BPA-free, this airtight container is ideal for transporting and securely storing products, whether liquid or solid in form.
  • Manufactured from translucent polypropylene
  • BPA-free
  • Microwave and dishwasher safe
  • Food contact safe
  • Temperature resistance (−25 to +100 °C)
This polypropylene box with an airtight lid is ideal for transporting and storing various products. Its lid features edges that allow safe and secure stacking. The box is resistant to shocks, chemicals, and temperature variations ranging from −25 to +100 °C.