Dispensing Can
Supplier: asecos
36488
MSGA36488EA 136 GBP
MSGA36488
Dispensing Can
Safety Cans
The dispensing can allows for safe, clean and comfortable dispensing of liquids in the workplace.
  • Resistant, chemically stable and UV-stable containers
  • 18 labels in 9 signal colours and a multilingual safety label for content labelling according to GHS
  • With stable handles for ergnomic handling
Includes 2 screw-on type outlets in different sizes.
Width: 173 mm
Height: 416 mm
Order Now
SPECIFICATIONS
Environmentally Preferable