Safety storage cabinets for chemicals, CS-Classic
Supplier: asecos
30495040 30495046 30495047 30495048 30495049 30495050
MSGA30495040EA 1390 GBP
MSGA30495040 MSGA30495046 MSGA30495047 MSGA30495048 MSGA30495049 MSGA30495050
Safety storage cabinets for chemicals, CS-Classic
Furniture Storage Cabinets
Storage of non-flammable chemicals and toxic substances in workroom.
  • Robust construction and longevity – body made of sheet steel with highgrade structured paint finish
  • No unauthorised use – doors lockable with cylinder locking (integration in a locking system possible)
  • Easy transport – integrated transport base with removable, optional base cover
  • Easy alignment – adjusting aids to compensate for uneven floor
  • Five more door colours available upon request at no extra cost
Certifications: TÜV approved.
Ordering information: Interior equipmentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable