USB Camera, Moticam® S20
Supplier: MOTIC
1100600101401
MOTI1100600101401EA 1470 GBP
MOTI1100600101401
USB Camera, Moticam® S20
Cameras
High Grade sCMOS USB Camera Moticam S20 for microscopy for an ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
Maximum details - If it is about maximum resolution for display or printing, if resolution in display or for printing is in your mind, Moticam S20 is your solution.
Ordering information: To be purchased separately - C-mount adapter not included to be purchased from the same brand of your microscope, compatible with your microscope model and fittingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Sensor size 1"
Frame rate 5472x3648 @ 18 fps, 2736x1824 @ 50 fps (max.)
Lens mount C-mount
Interfaces USB 3.1
Software Motic Images Plus 3.0 for Windows, OSX and Linux
Support device TWAIN, SDK and DirectShow Driver
Supported OS Windows 7/8/10, MAC OSX, Linux or higher