ω,4-Dibromoacetophenone

Supplier: Molekula
Danger

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

M87429637
MOLEM87429637EA 0 GBP
MOLEM87429637
ω,4-Dibromoacetophenone
ω,4-Dibromoacetophenone
Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Boiling Pt: 310
Melting Pt: 107…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000200
CAS Number: 99-73-0
EINECS: 202-783-6
UN: 3261
ADR: 8 II
Merck Index: 13,01412

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR