trans-4-(Aminomethyl)cyclohexanecarboxylic acid

Supplier: Molekula

Synonyms: HAKU, AMCA, TAMCHA, trans-4-(aminomethyl)cyclohexane-1-carboxylic acid, Tranexamic acid, AMCHA

M77717564
MOLEM77717564EA 0 GBP
MOLEM77717564
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD00001466
CAS Number: 1197-18-8
EINECS: 214-818-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR